siRNA Details
| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197940.2 | NP_922922.1 | 6 | 1364 | ||
| GenBank mRNA | BC057854.1 | - | 6 | 1153 | 6494304 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wipf2| Gene Symbol: Wipf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197940.2 | NP_922922.1 | 6 | 1364 | ||
| GenBank mRNA | BC057854.1 | - | 6 | 1153 | 6494304 | |