siRNA Details
| Gene Symbol: Pdrg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178939.2 | NP_849270.1 | 4 | 371 | ||
| GenBank mRNA | AK003410.1 | - | 4 | 373 | ||
| AK145939.1 | - | 4 | 416 | |||
| AY286302.1 | - | 4 | 352 | |||
| BC031699.1 | - | 4 | 371 | 4020218 | ||

Pdrg1
Genomic Map