siRNA Details
| Gene Symbol: Tmed10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026775.4 | NP_081051.1 | 2, 3 | 398 | ||
| GenBank mRNA | AK003676.1 | - | 2, 3 | 381 | ||
| AK084117.1 | - | 2, 3 | 381 | |||
| AK150605.1 | - | 2, 3 | 370 | |||
| AK152643.1 | - | 2, 3 | 381 | |||
| AK152656.1 | - | 2, 3 | 370 | |||
| AK186798.1 | - | 2, 3 | 143 | |||
| BC064755.1 | - | 2, 3 | 372 | 30029180 | ||
| BC085097.1 | - | 2, 3 | 377 | 5349826 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053467, X97443, AJ004912, FQ212056.

Tmed10
Genomic Map