siRNA Details
| Gene Symbol: Ppm1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176833.4 | NP_789803.1 | 5 | 825 | ||
| GenBank mRNA | AK016942.1 | - | 5 | 830 | ||
| AK154192.1 | - | 5 | 728 | |||
| AK164964.1 | - | 5 | 706 | |||
| BC042570.1 | - | 4 | 674 | 4241448 | ||

Ppm1f
Genomic Map