siRNA Details
| Gene Symbol: Letmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134093.2 | NP_598854.1 | 5 | 586 | ||
| GenBank mRNA | AF287293.1 | - | 5 | 526 | ||
| AF401483.1 | - | 5 | 419 | |||
| AK041312.1 | - | 4 | 915 | |||
| AK045256.1 | - | 5 | 406 | |||
| AK046496.1 | - | 4 | 390 | |||
| AK080221.1 | - | 5 | 506 | |||
| AK154676.1 | - | 5 | 501 | |||
| AK158718.1 | - | 4 | 363 | |||
| AK169341.1 | - | 5 | 506 | |||
| AK217327.1 | - | 3 | 130 | |||
| BC021361.1 | - | 2 | 258 | 5010892 | ||

Letmd1
Genomic Map