siRNA Details
| Gene Symbol: Cfap57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026789.4 | NP_081065.2 | 21 | 3495 | ||
| GenBank mRNA | AK003848.1 | - | 2 | 193 | ||
| BC062805.1 | - | 2 | 200 | 1478343 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap57| Gene Symbol: Cfap57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026789.4 | NP_081065.2 | 21 | 3495 | ||
| GenBank mRNA | AK003848.1 | - | 2 | 193 | ||
| BC062805.1 | - | 2 | 200 | 1478343 | ||