siRNA Details
| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026791.2 | NP_081067.2 | 9, 10 | 1307 | ||
| GenBank mRNA | AK003747.1 | - | 4, 5 | 371 | ||
| AK082085.1 | - | 8, 9 | 1796 | |||
| AK156709.1 | - | 8, 9 | 1457 | |||
| BC043658.1 | - | 9, 10 | 1268 | |||
| BC053694.1 | - | 7, 8 | 723 | |||
| BC060082.1 | - | 6, 7 | 538 | |||
| BC144773.1 | - | 9, 10 | 1673 | |||
| BC145661.1 | - | 9, 10 | 1673 | |||

Fbxw9
Genomic Map