siRNA Details
| Gene Symbol: Fbxw9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026791.2 | NP_081067.2 | 5 | 881 | ||
| GenBank mRNA | AK082085.1 | - | 4 | 1370 | ||
| AK156709.1 | - | 4 | 1031 | |||
| BC002276.1 | - | 1 | 399 | |||
| BC043658.1 | - | 5 | 842 | |||
| BC053694.1 | - | 3 | 297 | |||
| BC060082.1 | - | 2 | 112 | |||
| BC144773.1 | - | 5 | 1247 | |||
| BC145661.1 | - | 5 | 1247 | |||
| CF197934.1 | - | 3 | 285 | |||

Fbxw9
Genomic Map