siRNA Details
| Gene Symbol: Cryl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030004.3 | NP_084280.2 | 4 | 330 | ||
| GenBank mRNA | AB359906.1 | - | 4 | 295 | ||
| AF351609.1 | - | 4 | 375 | |||
| AK003913.1 | - | 4 | 368 | |||
| AK043569.1 | - | 4 | 377 | |||
| AK080625.1 | - | 4 | 351 | |||
| AK135834.1 | - | 4 | 344 | |||
| BC004074.1 | - | 4 | 330 | 3591359 | ||
| BC027064.1 | - | 4 | 347 | 5355916 | ||

Cryl1
Genomic Map