siRNA Details
| Gene Symbol: Cryl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030004.3 | NP_084280.2 | 8 | 910 | ||
| GenBank mRNA | AB359906.1 | - | 8 | 875 | ||
| AF351609.1 | - | 8 | 956 | |||
| AK003913.1 | - | 9 | 948 | |||
| AK043569.1 | - | 8 | 957 | |||
| AK080625.1 | - | 8 | 931 | |||
| AK135834.1 | - | 8 | 924 | |||
| BC004074.1 | - | 8 | 910 | 3591359 | ||
| BC027064.1 | - | 8 | 927 | 5355916 | ||

Cryl1
Genomic Map