siRNA Details
| Gene Symbol: Tmem216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277860.1 | NP_001264789.1 | 3, 4 | 243 | ||
| NM_001277861.1 | NP_001264790.1 | 3, 4 | 257 | |||
| NM_026798.3 | NP_081074.1 | 3, 4 | 316 | |||
| GenBank mRNA | AF332866.1 | - | 2, 3 | 182 | ||
| AK003740.1 | - | 3, 4 | 318 | |||
| AK013286.1 | - | 3, 4 | 247 | |||
| BC052053.1 | - | 3, 4 | 220 | 5693205 | ||
| BY127975.1 | - | 246 | ||||
| BY704000.1 | - | 3, 4 | 318 | |||
| DV063021.1 | - | 3, 4 | 311 | |||

Tmem216
Genomic Map