siRNA Details
| Gene Symbol: Tab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138667.3 | NP_619608.1 | 3 | 1375 | ||
| GenBank mRNA | AB093262.1 | - | 2 | 942 | ||
| AK041067.1 | - | 2 | 1900 | |||
| AK089164.1 | - | 3 | 1378 | |||
| AK147830.1 | - | 3 | 1186 | |||
| AY093701.1 | - | 2 | 1035 | |||
| BC004813.1 | - | 2 | 1042 | 3586132 | ||

Tab2
Genomic Map