siRNA Details
| Gene Symbol: Fndc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081416.1 | NP_001074885.1 | 12 | 4190 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CO432948.1 | - | 101 | Chr. - - [-] on Build | ||

Fndc1
Genomic Map