siRNA Details
| Gene Symbol: Scgb3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054037.2 | NP_473378.1 | 3 | 425 | ||
| NM_170727.2 | NP_733923.1 | 4 | 530 | |||
| GenBank mRNA | AF313456.1 | - | 3 | 410 | ||
| AF313457.1 | - | 4 | 515 | |||
| AK003988.1 | - | 3 | 412 | |||
| BC086910.1 | - | 3 | 407 | 6759656 | ||

Scgb3a1
Genomic Map