siRNA Details
| Gene Symbol: Scgb3a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054037.2 | NP_473378.1 | 2 | 136 | ||
| NM_170727.2 | NP_733923.1 | 3 | 241 | |||
| GenBank mRNA | AF313456.1 | - | 2 | 121 | ||
| AF313457.1 | - | 3 | 226 | |||
| AF436840.1 | - | 2 | 105 | |||
| AK003988.1 | - | 2 | 123 | |||
| AV578532.1 | - | 2 | 136 | |||
| BC086910.1 | - | 2 | 118 | 6759656 | ||

Scgb3a1
Genomic Map