siRNA Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026805.1 | NP_081081.1 | 16 | 1652 | ||
| GenBank mRNA | AK029670.1 | - | 15 | 1361 | ||
| AK047655.1 | - | 16 | 1645 | |||
| AK047764.1 | - | 16 | 1653 | |||
| AK053833.1 | - | 16 | 1643 | |||
| AK082744.1 | - | 16 | 1654 | |||
| BC059878.1 | - | 16 | 1605 | 6812082 | ||

Svop
Genomic Map