siRNA Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026805.1 | NP_081081.1 | 7, 8 | 828 | ||
| GenBank mRNA | AK029670.1 | - | 6, 7 | 537 | ||
| AK047655.1 | - | 7, 8 | 820 | |||
| AK047764.1 | - | 7, 8 | 829 | |||
| AK053833.1 | - | 7, 8 | 819 | |||
| AK082744.1 | - | 7, 8 | 830 | |||
| BC059878.1 | - | 7, 8 | 781 | 6812082 | ||

Svop
Genomic Map