siRNA Details
| Gene Symbol: Trpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175130.4 | NP_780339.2 | 11 | 1487 | ||
| GenBank mRNA | AJ575814.1 | - | 11 | 1466 | ||
| AK044094.1 | - | 1094 | ||||
| BC046472.1 | - | 1 | 159 | |||
| BC049165.1 | - | 9 | 1118 | |||
| BC058632.1 | - | 4 | 395 | |||
| BC096475.1 | - | 11 | 1463 | 4225167 | ||

Trpm4
Genomic Map