siRNA Details
| Gene Symbol: Trpm4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175130.4 | NP_780339.2 | 6 | 756 | ||
| GenBank mRNA | AJ575814.1 | - | 6 | 735 | ||
| AK044094.1 | - | 363 | ||||
| BC049165.1 | - | 4 | 387 | |||
| BC096475.1 | - | 6 | 732 | 4225167 | ||

Trpm4
Genomic Map