siRNA Details
Gene Symbol: Ppp1r27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026814.3 | NP_081090.1 | 1 | 181 | ||
GenBank mRNA | AK004064.1 | - | 1 | 182 | ||
BC115957.1 | - | 1 | 162 | 40055543 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
