siRNA Details
| Gene Symbol: Gtf2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026816.3 | NP_081092.1 | 6 | 528 | ||
| GenBank mRNA | AK051837.1 | - | 1 | 1463 | ||
| AK144858.1 | - | 6 | 529 | |||
| BC027173.1 | - | 6 | 524 | 3986466 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gtf2f2| Gene Symbol: Gtf2f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026816.3 | NP_081092.1 | 6 | 528 | ||
| GenBank mRNA | AK051837.1 | - | 1 | 1463 | ||
| AK144858.1 | - | 6 | 529 | |||
| BC027173.1 | - | 6 | 524 | 3986466 | ||