siRNA Details
| Gene Symbol: Rnf166 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033142.2 | NP_001028314.1 | 4 | 555 | ||
| GenBank mRNA | AK079883.1 | - | 4 | 537 | ||
| AK151813.1 | - | 4 | 555 | |||
| BC131924.1 | - | 4 | 554 | 40130201 | ||
| BC132281.1 | - | 4 | 554 | 40130558 | ||

Rnf166
Genomic Map