siRNA Details
| Gene Symbol: Lce1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026822.1 | NP_081098.1 | 2 | 588 | ||
| GenBank mRNA | AK003951.1 | - | 2 | 588 | ||
| BC069979.1 | - | 2 | 565 | |||
| BC082291.1 | - | 2 | 560 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lce1b| Gene Symbol: Lce1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026822.1 | NP_081098.1 | 2 | 588 | ||
| GenBank mRNA | AK003951.1 | - | 2 | 588 | ||
| BC069979.1 | - | 2 | 565 | |||
| BC082291.1 | - | 2 | 560 | |||