siRNA Details
| Gene Symbol: 1110032F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167996.1 | NP_001161468.1 | 1 | 1641 | ||
| GenBank mRNA | AK082199.1 | - | 1 | 1386 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1110032F04Rik| Gene Symbol: 1110032F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167996.1 | NP_001161468.1 | 1 | 1641 | ||
| GenBank mRNA | AK082199.1 | - | 1 | 1386 | ||