siRNA Details
| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005417.2 | NP_001005417.1 | 7 | 1224 | ||
| NM_003780.4 | NP_003771.1 | 7 | 1424 | |||
| NM_030587.2 | NP_085076.2 | 7 | 1225 | |||
| XM_017002716.1 | XP_016858205.1 | 7 | 1182 | |||
| XM_017002717.1 | XP_016858206.1 | 7 | 1385 | |||
| GenBank mRNA | AB024434.1 | - | 6 | 1070 | ||
| AF038660.1 | - | 6 | 1114 | |||
| AK074533.1 | - | 5 | 722 | |||
| AK095873.1 | - | 7 | 1852 | |||
| AK293418.1 | - | 7 | 1174 | |||
| AL137647.1 | - | 5 | 619 | |||
| BC096821.1 | - | 7 | 1192 | 30558762 | ||
| Y12510.1 | - | 6 | 1062 | |||

B4GALT2
Genomic Map