siRNA Details
| Gene Symbol: Rbfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199197.1 | NP_954667.1 | 4 | 614 | ||
| GenBank mRNA | AK004019.1 | - | 4 | 633 | ||
| AK145807.1 | - | 4 | 653 | |||
| BC064066.1 | - | 4 | 614 | 935634 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rbfa| Gene Symbol: Rbfa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199197.1 | NP_954667.1 | 4 | 614 | ||
| GenBank mRNA | AK004019.1 | - | 4 | 633 | ||
| AK145807.1 | - | 4 | 653 | |||
| BC064066.1 | - | 4 | 614 | 935634 | ||