siRNA Details
| Gene Symbol: Lrrc16a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026825.3 | NP_081101.3 | 19 | 1640 | ||
| GenBank mRNA | AK043591.1 | - | 4 | 220 | ||
| AK049059.1 | - | 1 | 16 | |||
| AY437876.1 | - | 19 | 1645 | |||
| BC150819.1 | - | 18 | 1554 | |||

Lrrc16a
Genomic Map