siRNA Details
| Gene Symbol: Smek1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160214.1 | NP_001153686.1 | 10 | 2403 | ||
| NM_211355.2 | NP_997594.1 | 10 | 2364 | |||
| GenBank mRNA | AK051239.1 | - | 10 | 2289 | ||
| AK148868.1 | - | 3 | 236 | |||
| AK152480.1 | - | 10 | 2400 | |||
| AK156272.1 | - | 10 | 2326 | |||
| AK173331.1 | - | 9 | 4072 | |||
| BC064465.1 | - | 10 | 2062 | 6825400 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001271174, NM_001108050, EV776435.

Smek1
Genomic Map