siRNA Details
| Gene Symbol: Krtap22-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191018.1 | NP_001177947.1 | 1 | 325 | ||
| GenBank mRNA | AA763224.1 | - | 1 | 303 | ||
| AK004025.1 | - | 1 | 326 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap22-2| Gene Symbol: Krtap22-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191018.1 | NP_001177947.1 | 1 | 325 | ||
| GenBank mRNA | AA763224.1 | - | 1 | 303 | ||
| AK004025.1 | - | 1 | 326 | |||