siRNA Details
| Gene Symbol: Anln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028390.3 | NP_082666.2 | 12, 13 | 2375 | ||
| GenBank mRNA | AK045703.1 | - | 9, 10 | 1339 | ||
| AK046102.1 | - | 1, 2 | 161 | |||
| BC030489.1 | - | 5, 6 | 717 | 5373071 | ||
| BC032164.1 | - | 12, 13 | 2393 | 5346703 | ||

Anln
Genomic Map