siRNA Details
| Gene Symbol: Rreb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039188.1 | NP_001034277.1 | 10 | 3849 | ||
| NM_001177868.1 | NP_001171339.1 | 10 | 3574 | |||
| NM_001177869.1 | NP_001171340.1 | 9 | 3510 | |||
| NM_026830.2 | NP_081106.1 | 10 | 3817 | |||
| GenBank mRNA | AK147260.1 | - | 10 | 4972 | ||
| AK147653.1 | - | 10 | 4972 | |||
| AK154980.1 | - | 10 | 3817 | |||
| AK170652.1 | - | 10 | 3853 | |||
| AK171375.1 | - | 10 | 3554 | |||
| AY946044.1 | - | 7 | 3201 | |||
| BC023785.1 | - | 1 | 785 | |||
| BC080680.1 | - | 10 | 3663 | 6330949 | ||

Rreb1
Genomic Map