siRNA Details
| Gene Symbol: Cgrrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026832.3 | NP_081108.2 | 5 | 652 | ||
| GenBank mRNA | AK030755.1 | - | 5 | 654 | ||
| AK154797.1 | - | 5 | 728 | |||
| BC037677.1 | - | 5 | 668 | 3491666 | ||
| BC114536.1 | - | 4 | 490 | |||

Cgrrf1
Genomic Map