siRNA Details
| Gene Symbol: Synpo2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175132.3 | NP_780341.1 | 4 | 2559 | ||
| GenBank mRNA | AK053054.1 | - | 4 | 2559 | ||
| AK084541.1 | - | 2 | 1846 | |||
| BC137972.1 | - | 4 | 2530 | 8860991 | ||
| BC145070.1 | - | 4 | 2529 | |||

Synpo2l
Genomic Map