siRNA Details
| Gene Symbol: Krtap4-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026834.2 | NP_081110.1 | 1 | 699 | ||
| GenBank mRNA | AK004258.1 | - | 1 | 700 | ||
| BC120646.1 | - | 1 | 666 | |||
| BC120648.1 | - | 1 | 666 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap4-6| Gene Symbol: Krtap4-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026834.2 | NP_081110.1 | 1 | 699 | ||
| GenBank mRNA | AK004258.1 | - | 1 | 700 | ||
| BC120646.1 | - | 1 | 666 | |||
| BC120648.1 | - | 1 | 666 | |||