siRNA Details
| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285812.1 | NP_001272741.1 | 2, 3 | 331 | ||
| NM_001285814.1 | NP_001272743.1 | 3, 4 | 396 | |||
| NM_001285815.1 | NP_001272744.1 | 3, 4 | 414 | |||
| NM_026837.3 | NP_081113.1 | 2, 3 | 310 | |||
| NM_029589.1 | NP_083865.1 | 3, 4 | 232 | |||
| GenBank mRNA | AK004164.1 | - | 2, 3 | 273 | ||
| AK005104.1 | - | 2, 3 | 217 | |||
| AK005885.1 | - | 3, 4 | 236 | |||
| BC019937.1 | - | 2, 3 | 236 | 4023311 | ||
| BC020121.1 | - | 2, 3 | 240 | 3991677 | ||
| BC039805.1 | - | 1, 2 | 615 | |||

Tmem53
Genomic Map