siRNA Details
| Gene Symbol: Gucd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175133.1 | NP_780342.1 | 6 | 802 | ||
| GenBank mRNA | AK034509.1 | - | 686 | |||
| AK088820.1 | - | 6 | 747 | |||
| AK157404.1 | - | 6 | 861 | |||
| AK170872.1 | - | 6 | 838 | |||
| BC024851.1 | - | 6 | 756 | 5359071 | ||
| BC043463.1 | - | 6 | 802 | 5042988 | ||
| BC118613.1 | - | 6 | 860 | 40107160 | ||
| BC119795.1 | - | 6 | 860 | 40107164 | ||

Gucd1
Genomic Map