siRNA Details
| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037622.1 | - | 8 | 1307 | ||
| BC172146.1 | - | 8 | 1055 | |||
| DQ924536.1 | - | 8 | 1366 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubr3| Gene Symbol: Ubr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK037622.1 | - | 8 | 1307 | ||
| BC172146.1 | - | 8 | 1055 | |||
| DQ924536.1 | - | 8 | 1366 | |||