siRNA Details
| Gene Symbol: Tmem214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144525.3 | NP_653108.2 | 6 | 786 | ||
| GenBank mRNA | AK034883.1 | - | 6 | 788 | ||
| AK053521.1 | - | 802 | ||||
| AK143078.1 | - | 788 | ||||
| AK165285.1 | - | 5 | 949 | |||
| AK166510.1 | - | 6 | 781 | |||
| BC022142.1 | - | 2 | 63 | 5101205 | ||
| BC026651.1 | - | 6 | 702 | 4987771 | ||
| BC029150.1 | - | 6 | 774 | 4954376 | ||

Tmem214
Genomic Map