siRNA Details
| Gene Symbol: Tmem214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144525.3 | NP_653108.2 | 1 | 151 | ||
| GenBank mRNA | AK034883.1 | - | 1 | 153 | ||
| AK053521.1 | - | 167 | ||||
| AK143078.1 | - | 153 | ||||
| AK165285.1 | - | 1 | 163 | |||
| AK166510.1 | - | 1 | 146 | |||
| BC026651.1 | - | 1 | 67 | 4987771 | ||
| BC029150.1 | - | 1 | 139 | 4954376 | ||

Tmem214
Genomic Map