siRNA Details
| Gene Symbol: Prr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026841.1 | NP_081117.1 | 2 | 364 | ||
| GenBank mRNA | AK004321.1 | - | 1 | 219 | ||
| BC060289.1 | - | 2 | 364 | |||
| BC094615.1 | - | 2 | 332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prr32| Gene Symbol: Prr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026841.1 | NP_081117.1 | 2 | 364 | ||
| GenBank mRNA | AK004321.1 | - | 1 | 219 | ||
| BC060289.1 | - | 2 | 364 | |||
| BC094615.1 | - | 2 | 332 | |||