siRNA Details
| Gene Symbol: Dock5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177780.3 | NP_808448.2 | 26 | 2916 | ||
| GenBank mRNA | AK147975.1 | - | 26 | 2778 | ||
| BC050992.1 | - | 2 | 114 | |||
| BC158046.1 | - | 26 | 2776 | 9007351 | ||
| BC158135.1 | - | 26 | 2776 | 9088200 | ||
| BC158138.1 | - | 26 | 2776 | 9088202 | ||
| CF746784.1 | - | 7 | 627 | |||

Dock5
Genomic Map