siRNA Details
| Gene Symbol: Btbd10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133700.2 | NP_598461.1 | 11 | 1591 | ||
| GenBank mRNA | BC002151.1 | - | 2 | 211 | ||
| BC038876.1 | - | 2 | 157 | |||
| BC039798.1 | - | 1 | 507 | |||
| BC050781.1 | - | 11 | 1591 | 6591397 | ||

Btbd10
Genomic Map