siRNA Details
Gene Symbol: Ddi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017966.2 | NP_001017966.1 | 8 | 1361 | ||
GenBank mRNA | AK129015.1 | - | 1339 | |||
BC021415.1 | - | 8 | 1342 | |||
BC031491.1 | - | 1 | 115 | |||
CD550969.1 | - | 3 | 273 |

More Information
Other Species Targeted

Panther Classification:
