siRNA Details
| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080932.2 | NP_001074401.2 | 5 | 1061 | ||
| GenBank mRNA | AK078027.1 | - | 1 | 69 | ||
| AK150439.1 | - | 5 | 715 | |||
| EU882160.1 | - | 5 | 1061 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxk2| Gene Symbol: Foxk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080932.2 | NP_001074401.2 | 5 | 1061 | ||
| GenBank mRNA | AK078027.1 | - | 1 | 69 | ||
| AK150439.1 | - | 5 | 715 | |||
| EU882160.1 | - | 5 | 1061 | |||