siRNA Details
| Gene Symbol: Ankrd46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175134.4 | NP_780343.1 | 3 | 324 | ||
| GenBank mRNA | AB076382.1 | - | 3 | 319 | ||
| AK088304.1 | - | 3 | 285 | |||
| AK148049.1 | - | 3 | 285 | |||
| AK186948.1 | - | 1 | 83 | |||
| BC035553.1 | - | 3 | 325 | |||
| BC086659.1 | - | 3 | 291 | 30607272 | ||
| BC094595.1 | - | 3 | 287 | 5707829 | ||
| BQ715954.1 | - | 3 | 324 | |||

Ankrd46
Genomic Map