siRNA Details
| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103181.1 | NP_001096651.1 | 8 | 1260 | ||
| NM_054040.3 | NP_473381.1 | 8 | 2102 | |||
| GenBank mRNA | AF219945.1 | - | 8 | 1506 | ||
| AK147643.1 | - | 8 | 1200 | |||
| AK173165.1 | - | 8 | 2102 | |||
| BC138024.1 | - | 8 | 1437 | 8861043 | ||
| BC138025.1 | - | 8 | 1437 | 8861044 | ||

Tulp4
Genomic Map