siRNA Details
| Gene Symbol: Rnf208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176834.2 | NP_789804.2 | 1 | 473 | ||
| GenBank mRNA | BC030073.1 | - | 2 | 275 | 5254639 | |
| CX206020.1 | - | 1 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf208| Gene Symbol: Rnf208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176834.2 | NP_789804.2 | 1 | 473 | ||
| GenBank mRNA | BC030073.1 | - | 2 | 275 | 5254639 | |
| CX206020.1 | - | 1 | 473 | |||