siRNA Details
| Gene Symbol: Dtwd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026854.3 | NP_081130.1 | 4 | 503 | ||
| GenBank mRNA | AK004450.1 | - | 4 | 478 | ||
| AK144689.1 | - | 4 | 478 | |||
| BC075693.1 | - | 6 | 597 | |||
| BC139347.1 | - | 4 | 469 | 8862369 | ||
| BC139348.1 | - | 4 | 469 | 8862370 | ||

Dtwd2
Genomic Map