siRNA Details
| Gene Symbol: 1190002N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033145.2 | NP_001028317.2 | 3 | 1180 | ||
| GenBank mRNA | AK162197.1 | - | 2 | 304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1190002N15Rik| Gene Symbol: 1190002N15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033145.2 | NP_001028317.2 | 3 | 1180 | ||
| GenBank mRNA | AK162197.1 | - | 2 | 304 | ||