siRNA Details
| Gene Symbol: Arv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026855.4 | NP_081131.1 | 2 | 256 | ||
| GenBank mRNA | AK004398.1 | - | 2 | 257 | ||
| BC119376.1 | - | 2 | 236 | 8734125 | ||
| BC120631.1 | - | 2 | 236 | 40129554 | ||
| BC144875.1 | - | 2 | 236 | |||
| BC144876.1 | - | 2 | 236 | |||

Arv1
Genomic Map